Learn More
Med Vet International Miltex Dingman Periosteal Elevator, Double-Ended, 7.25in, German, Pm-4770
Supplier:  Med Vet International MILPM4770
Encompass_Preferred
The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. This is a Miltex Dingman Periosteal Elevator. Elevators are instruments used in dental extractions or surgical procedures. In dental procedures they may be used to loosen teeth prior to forcep extraction, to remove roots or impacted teeth, when teeth are compromised and susceptible to fracture or when they are malpositioned and cannot be reached with forceps. In surgical procedures they can be used for scraping, elevating or dissecting bones or tissues. They are made from premium-grade German stainless steel. Specifications: Double-Ended, 7. 25in, Width 1 - 6. 2 mm Wide, Width 2 - 7. 2 mm Wide, PM-4770
Catalog No. 50-282-9296