Learn More
Medchemexpress LLC Elevenostat | 1454902-97-6 | 95.1% | C16H17N3O4 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14575750MG
Encompass_Preferred
Elevenostat is a selective HDAC11 inhibitor that induces apoptosis in multiple myeloma cells and demonstrates an anti-tumor effect. It also inhibits the maturation of mouse oocytes and is intended for research use only.
  • Selective HDAC11 inhibitor (IC50 of 0.235 μM)
  • Induces apoptosis of multiple myeloma cells
  • Demonstrates anti-tumor effect
  • Inhibits maturation of mouse oocytes
  • For research use only
Catalog No. 50-003-56271