Learn More
Medchemexpress LLC HY-111772 1mg , Elexacaftor CAS:2216712-66-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1117721MG
Encompass_Preferred
HY-111772 1mg Elexacaftor CAS: 2216712-66-0 Elexacaftor (VX-445, Compound 1) is a modulator of cystic fibrosis transmembrane conductance regulator (CFTR). Elexacaftor (VX-445, Compound 1) facilitates the processing and trafficking of CFTR to increase the amount of CFTR at the cell surface. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0534