Learn More
Medchemexpress LLC Elexacaftor | 2216712-66-0 | 597.65 | 10 MM 1 ML
SDP
Supplier:  Medchemexpress LLC HY11177210MM/1ML
Encompass_Preferred
Elexacaftor (VX-445, Compound 1) is a modulator of cystic fibrosis transmembrane conductance regulator (CFTR). It facilitates the processing and trafficking of CFTR to increase the amount of CFTR at the cell surface. This next-generation CFTR corrector is designed to restore Phe508del CFTR protein function and has the potential to treat cystic fibrosis.
  • Orally active and highly potent CFTR corrector.
  • Restores processing and trafficking of F508del CFTR protein to the cell surface in human bronchial epithelial (HBE) cells.
  • Binds to F508del CFTR protein and acts as a proteostasis corrector.
  • Can be used for cystic fibrosis research.
Catalog No. 50-004-40012