Learn More
Selleck Chemical LLC Elexacaftor (VX-445) 2mg 2216712-66-0
SDP
Supplier:  Selleck Chemical LLC S88512MG
Encompass_Preferred
Elexacaftor (VX-445) is a next-generation cystic fibrosis transmembrane conductance regulator (CFTR) corrector.This is a compound which is not chiral purity. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1761795