Learn More
TARGETMOL CHEMICALS INC ELIGLUSTAT 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T366310MG
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Eliglustat (Genz 99067) is an oral inhibitor of glucosylceramide synthase which is used in the therapy of type 1 Gaucher disease. purity: 99%
Catalog No. 50-257-6892