Learn More
Selleck Chemical LLC Eliglustat 5mg
Supplier: Selleck Chemical LLC S78525MG
Eliglustat (GENZ-112638) inhibits glucosylceramide synthase(GCS) (IC50=20 nM in intact MDCK cells), thus reducing the load of glucosylceramide influx into the lysosome. *For Research & Development use only. Product is not intended for drug, household, or human consumption.