Learn More
Selleck Chemical LLC Elimusertib S9864-1G
SDP
Supplier:  Selleck Chemical LLC S98641G
Encompass_Preferred
Elimusertib (BAY-1895344) is a very potent and highly selective ATR (ataxia telangiectasia and Rad3-related) inhibitor with IC50 of 7 nM Elimusertib potently inhibits proliferation of a broad spectrum of human tumor cell lines with median IC50 of 78 nM
Catalog No. 50-000-24685