Learn More
Medchemexpress LLC Elimusertib hydrochloride | 99.9% | 411.89 | 100 MG
SDP
Supplier:  Medchemexpress LLC 100MGHY101566A
Encompass_Preferred
Elimusertib hydrochloride (BAY 1895344) is a potent, orally active, and selective ATR inhibitor with an IC50 of 7 nM. It demonstrates anti-tumor activity and can be used for research on solid tumors and lymphomas.
  • Potent, orally active, and selective ATR inhibitor
  • Demonstrates anti-tumor activity
  • Applicable for research on solid tumors and lymphomas
  • Inhibits proliferation of human tumor cell lines
  • Suppresses hydroxyurea-induced H2AX phosphorylation
  • Shows good selectivity against related kinases
  • Potent antiproliferative activity against various cancer cell lines in vitro
  • Potent anti-tumor efficacy in monotherapy in xenograft models
  • Causes complete tumor remission in mantle cell lymphoma models
  • Synergistic antitumor activity with Carboplatin in relevant models
  • Exhibits moderate oral bioavailability
Catalog No. 50-003-64822