Learn More
BioVendor, LLC Adiponectin HMW (high molecular weight) Mouse/Rat ELISA (96 wells)
SDP
Supplier:  BioVendor, LLC 29790701
Encompass
TypeSandwich ELISA, HRP-labelled antibodyApplicationsSerumSample Requirements10 µl/wellShippingAt ambient temperature. Upon receipt, store the product at the temperature recommended below.Storage/ExpirationStore the complete kit at 2–8°C. Under these conditions, the kit is stable until the expiration date (see label on the box). Calibration Range3.13–200 ng/ml (In the case of 50X dilution, original HMW adiponectin range is 156–10 000 ng/ml)Limit of Detection0.313 ng/ml
Catalog No. 50-893-002