Learn More
BioVendor, LLC Apo B-48 Human ELISA (96 wells)
SDP
Supplier:  BioVendor, LLC 29888901
Encompass
TypeSandwich ELISA, Biotin-labelled antibodyApplicationsSerum, Plasma-EDTA, Plasma-HeparinSample Requirements50 µl/well (Diluted to 100X with buffer solution ©)ShippingAt ambient temperature. Upon receipt, store the product at the temperature recommended below.Storage/ExpirationStore the complete kit at 2–8°C. Under these conditions, the kit is stable until the expiration date (see label on the box). Calibration Range2.5–160 ng/mlLimit of Detection0.198 ng/ml
Catalog No. 50-893-021