Learn More
BioVendor, LLC C-Peptide (U-type) Mouse ELISA (96 wells)
SDP
Supplier:  BioVendor, LLC 29990901
Encompass
C-Peptide (U-type) Mouse ELISA (96 wells). Immunoassay kit to quantify total C-Peptide in mouse serum, plasma, or cell culture supernatant. The kit recognizes the common sequences between C-peptide 1 and 2, allowing measurement of the total amount of C-peptide. SAMPLE REQ: 10 l/well. CAL. RANGE: 303000 pg/ml. DETECTION LIMIT: 1.5 pg/ml. SUMMARY: Mouse C-peptide 1 is a single chain peptide composed of 29 amino acids, while C-peptide 2 is composed of 31 residues. Biological half life of C-peptide is several times longer than that of insulin. Measurement of C-peptide is useful in estimation of pancreatic function for insulin synthesis and secretion. Urinary C-peptide concentration is well correlated to its blood level. C-peptide measurement is also useful in estimation of insulin secretion by cultured islet of Langerhans because very often insulin is added to the culture medium, and it is difficult to discriminate secreted insulin from added insulin. By FUJIFILM Wako Shibayagi Corp.
Catalog No. 50-893-008