Learn More
BioVendor, LLC ELISA CLUSTERIN HUMAN 96W
SDP
Supplier:  BioVendor, LLC RD194034200R
Encompass
TypeSandwich ELISA, Biotin-labelled antibodyApplicationsSerum, Plasma-EDTA, Plasma-Heparin, Plasma-Citrate, Urine, Cerebrospinal fluidSample Requirements5 µl/wellShippingAt ambient temperature. Upon receipt, store the product at the temperature recommended below.Storage/ExpirationStore the complete kit at 2–8°C. Under these conditions, the kit is stable until the expiration date (see label on the box). Calibration Range5–160 ng/mlLimit of Detection0.5 ng/ml
Catalog No. 50-894-207