Learn More
BioVendor, LLC ELISA CYSTATIN C MOUSE 96W
SDP
Supplier:  BioVendor, LLC RD291009200R
Encompass
TypeSandwich ELISA, Biotin-labelled antibodyApplicationsSerumSample Requirements5 µl/wellShippingAt ambient temperature. Upon receipt, store the product at the temperature recommended below.Storage/ExpirationStore the complete kit at 2–8°C. Under these conditions, the kit is stable until the expiration date (see label on the box). Calibration Range0.156–10 ng/mlLimit of Detection0.04 ng/ml
Catalog No. 50-894-211