Learn More
BioVendor, LLC FIBROBLAST GROWTH FACTOR 19 HUMAN ELISA 96W
SDP
Supplier:  BioVendor, LLC RD191107200R
Encompass
TypeSandwich ELISA, Biotin-labelled antibodyApplicationsSerum, Plasma-EDTA, Plasma-Heparin, Plasma-CitrateSample Requirements50 µl/wellShippingAt ambient temperature. Upon receipt, store the product at the temperature recommended below.Storage/ExpirationStore the complete kit at 2–8°C. Under these conditions, the kit is stable until the expiration date (see label on the box). Calibration Range12.5–800 pg/mlLimit of Detection4.8 pg/ml
Catalog No. 50-894-215