Learn More
BioVendor, LLC FIBROBLAST GROWTH FACTOR 21 HUMAN ELISA 96W
SDP
Supplier:  BioVendor, LLC RD191108200R
Encompass
TypeSandwich ELISA, Biotin-labelled antibodyApplicationsSerum, PlasmaSample Requirements75 µl/wellShippingAt ambient temperature. Upon receipt, store the product at the temperature recommended below.Storage/ExpirationStore the complete kit at 2–8°C. Under this condition, the kit is stable until the expiration date (see the label on the box). Calibration Range30–1920 pg/mlLimit of Detection7 pg/ml
Catalog No. 50-894-216