Learn More
BioVendor, LLC ELISA IMMUNOGLOB. FLC HUMAN (96 wells)
SDP
Supplier:  BioVendor, LLC RD194088100R
Encompass
TypeSandwich ELISA, Biotin-labelled antibodyApplicationsSerum, Plasma-EDTA, Plasma-Heparin, Plasma-Citrate, UrineSample Requirements2×5 µl/wellCalibration Range10–320 µg/l (FLC kappa)17.5–560 µg/l (FLC lambda)Limit of DetectionFLC kappa ELISA Limit of detection (LOD) (defined as concentration of analyte giving absorbance higher than mean absorbance of blank plus three standard deviations of the absorbance of blank: Ablank + 3×SDblank) is calculated from the real FLC kappa values in wells and is 6 μg/L.FLC lambda ELISA Limit of detection (LOD) (defined as concentration of analyte giving absorbance higher than mean absorbance of blank plus three standard deviations of the absorbance of blank: Ablank + 3×SDblank) is calculated from the real FLC lambda values in wells and is 5 μg/L.
Catalog No. 50-894-219