Learn More
Biomatik Corporation Phospho-VEGFR2 (Tyr951) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51592
Encompass_Preferred
Phospho-VEGFR2 (Tyr951) Colorimetric Cell-Based ELISA Kit for detecting Phospho-VEGFR2 (Tyr951) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P35968. Gene ID: 3791. Target Synonyms: FLK1; KDR; kinase insert domain receptor; protein-tyrosine kinase receptor Flk-1; vascular endothelial growth factor receptor 2; VGFR2; VGR2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9084