Learn More
Biomatik Corporation Phospho-SHP-1 (Tyr564) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51611
Encompass_Preferred
Phospho-SHP-1 (Tyr564) Colorimetric Cell-Based ELISA Kit for detecting Phospho-SHP-1 (Tyr564) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P29350. Gene ID: 5777. Target Synonyms: 70Z-SHP; HCP; HCPH; Hematopoietic cell protein-tyrosine phosphatase; Protein-tyrosine phosphatase 1C; Protein-tyrosine phosphatase SHP-1; Protein-tyrosine phosphatase; non-receptor type 6; PTN6; PTP-1C; PTP1C; PTPN6; SH-PTP1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9086