Learn More
Biomatik Corporation Phospho-Tyrosine Hydroxylase (Ser19) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50997
Encompass_Preferred
Phospho-Tyrosine Hydroxylase (Ser19) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Tyrosine Hydroxylase (Ser19) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P07101. Gene ID: 7054. Target Synonyms: TH isoform 3; TH isoform a; TH-4; TY3H; TYH; Tyrosine 3-hydroxylase; Tyrosine 3-monooxygenase; tyrosine hydroxylase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1209