Learn More
Biomatik Corporation Phospho-eEF2K (Ser366) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51020
Encompass_Preferred
Phospho-eEF2K (Ser366) Colorimetric Cell-Based ELISA Kit for detecting Phospho-eEF2K (Ser366) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: O00418. Gene ID: 29904. Target Synonyms: Calcium/calmodulin-dependent eukaryotic elongation factor-2 kinase; EF2K; Elongation factor 2 kinase; eEF-2 kinase; eEF-2K; kinase eEF2K. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1226