Learn More
Biomatik Corporation Phospho-SEPARASE (Ser801) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51034
Encompass_Preferred
Phospho-SEPARASE (Ser801) Colorimetric Cell-Based ELISA Kit for detecting Phospho-SEPARASE (Ser801) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q14674. Gene ID: 9700. Target Synonyms: Separin; Caspase-like protein ESPL1; Extra spindle poles-like 1 protein; Separase; ESPL1; ESP1; KIAA0165. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1239