Learn More
Biomatik Corporation Phospho-E2F1 (Thr433) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51063
Encompass_Preferred
Phospho-E2F1 (Thr433) Colorimetric Cell-Based ELISA Kit for detecting Phospho-E2F1 (Thr433) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q01094. Gene ID: 1869. Target Synonyms: E2F-1; PBR3; PRB-binding protein E2F-1; RBAP-1; RBBP-3; RBBP3; Retinoblastoma binding protein 3; Retinoblastoma-associated protein 1; Transcription factor E2F1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1264