Learn More
Biomatik Corporation Phospho-MYPT1 (Thr853) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51126
Encompass_Preferred
Phospho-MYPT1 (Thr853) Colorimetric Cell-Based ELISA Kit for detecting Phospho-MYPT1 (Thr853) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: O14974. Gene ID: 4659. Target Synonyms: MBS; Myosin phosphatase target subunit 1; PPP1R12A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9017