Learn More
Biomatik Corporation Phospho-Histone H3 (Ser28) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51072
Encompass_Preferred
Phospho-Histone H3 (Ser28) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Histone H3 (Ser28) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P68431; Q71DI3; P84243. Gene ID: 8350; 8351; 8352; 8353; 8354; 8355; 8356; 8357; 8358; 8968. Target Synonyms: H3/a; H3/c; H3/d; H3/f; H3/h; H3/i; H3/j; H3/k; H3/l; H31; HIST1H3A; histone H3.1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1272