Learn More
Biomatik Corporation Phospho-IκB-β (Thr19) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51075
Encompass_Preferred
Phospho-IκB-β (Thr19) Colorimetric Cell-Based ELISA Kit for detecting Phospho-IκB-β (Thr19) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q15653. Gene ID: 4793. Target Synonyms: I-kappa-B-beta; IKB-B; IKBB; IkappaBbeta; NF-kappa-BIB; NF-kappaB inhibitor beta; NFKBIB; TR- interacting protein 9; TRIP9; Thyroid receptor interacting protein 9. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1275