Learn More
Biomatik Corporation Phospho-Lamin A/C (Ser392) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51118
Encompass_Preferred
Phospho-Lamin A/C (Ser392) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Lamin A/C (Ser392) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P02545. Gene ID: 4000. Target Synonyms: 70 kDa lamin; LAMA; LMN1; LMNA. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1313