Learn More
Biomatik Corporation Phospho-PDE4D (Ser190/53) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51140
Encompass_Preferred
Phospho-PDE4D (Ser190/53) Colorimetric Cell-Based ELISA Kit for detecting Phospho-PDE4D (Ser190/53) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q08499. Gene ID: 5144. Target Synonyms: DPDE3; PDE43; PDE4D4; cAMP-specific 3',5'-cyclic phosphodiesterase 4D. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1330