Learn More
Biomatik Corporation Phospho-Progesterone Receptor (Ser400) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51149
Encompass_Preferred
Phospho-Progesterone Receptor (Ser400) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Progesterone Receptor (Ser400) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P06401. Gene ID: 5241. Target Synonyms: NR3C3; nuclear receptor 3C3; PGR; PRGR; PR. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1336