Learn More
Biomatik Corporation Phospho-SMC1 (Ser966) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51156
Encompass_Preferred
Phospho-SMC1 (Ser966) Colorimetric Cell-Based ELISA Kit for detecting Phospho-SMC1 (Ser966) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q14683. Gene ID: 8243. Target Synonyms: Chromosome segregation protein SmcB; DXS423E; KIAA0178; SB1.8/DXS423E protein; SM1A; SMC1A; SMC1L1; SMC1alpha protein; Sb1.8; Structural maintenance of chromosome 1-like 1 protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1343