Learn More
Biomatik Corporation Phospho-TEBP (Ser113) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51163
Encompass_Preferred
Phospho-TEBP (Ser113) Colorimetric Cell-Based ELISA Kit for detecting Phospho-TEBP (Ser113) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q15185. Gene ID: 10728. Target Synonyms: Prostaglandin E synthase 3; Cytosolic prostaglandin E2 synthase; cPGES; Telomerase-binding protein p23; Hsp90 co-chaperone; Progesterone receptor complex p23; PTGES3P23; TEBP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1349