Learn More
Biomatik Corporation Phospho-ACK1 (Tyr284) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51175
Encompass_Preferred
Phospho-ACK1 (Tyr284) Colorimetric Cell-Based ELISA Kit for detecting Phospho-ACK1 (Tyr284) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q07912. Gene ID: 10188. Target Synonyms: ACK1; Activated p21cdc42Hs kinase; Non-receptor protein tyrosine kinase Ack; TNK2; kinase ACK1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9027