Learn More
Biomatik Corporation Phospho-ETK (Tyr40) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51190
Encompass_Preferred
Phospho-ETK (Tyr40) Colorimetric Cell-Based ELISA Kit for detecting Phospho-ETK (Tyr40) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P51813. Gene ID: 660. Target Synonyms: BMX; BMX NON-receptor tyrosine kinase; Bone marrow kinase BMX; Cytoplasmic BMX; Epithelial and endothelial tyrosine kinase; NTK38; Protein tyrosine kinase BMX; kinase Etk. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1371