Learn More
Biomatik Corporation Phospho-PBK/TOPK (Thr9) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51208
Encompass_Preferred
Phospho-PBK/TOPK (Thr9) Colorimetric Cell-Based ELISA Kit for detecting Phospho-PBK/TOPK (Thr9) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q96KB5. Gene ID: 55872. Target Synonyms: MPAKK-like protein kinase cap site form 2; NORI-3; TOPK. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1386