Learn More
Biomatik Corporation Phospho-53BP1 (Ser6) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51222
Encompass_Preferred
Phospho-53BP1 (Ser6) Colorimetric Cell-Based ELISA Kit for detecting Phospho-53BP1 (Ser6) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q12888. Gene ID: 7158. Target Synonyms: p53-binding protein 1; P531; TP53B; TP53BP1; TRP53BP1; Tumor suppressor p53-binding protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1397