Learn More
Biomatik Corporation Phospho-ATRIP (Ser68) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51228
Encompass_Preferred
Phospho-ATRIP (Ser68) Colorimetric Cell-Based ELISA Kit for detecting Phospho-ATRIP (Ser68) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q8WXE1. Gene ID: 84126. Target Synonyms: ATIP; ATM and Rad3 related interacting protein; ATR-interacting protein; TREX1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1402