Learn More
Biomatik Corporation Phospho-Breast Tumor Kinase (Tyr447) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51232
Encompass_Preferred
Phospho-Breast Tumor Kinase (Tyr447) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Breast Tumor Kinase (Tyr447) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q13882. Gene ID: 5753. Target Synonyms: Breast tumor kinase; PTK6; Tyrosine-protein kinase 6; Tyrosine-protein kinase brk. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1406