Learn More
Biomatik Corporation Phospho-PTP1B (Ser50) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51247
Encompass_Preferred
Phospho-PTP1B (Ser50) Colorimetric Cell-Based ELISA Kit for detecting Phospho-PTP1B (Ser50) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P18031. Gene ID: 5770. Target Synonyms: CPTP1; Protein-protein-tyrosine phosphatase HA2; Protein-tyrosine phosphatase 1B; Protein-tyrosine phosphatase; non-receptor type 1; PTN1; PTP-1B; PTP-HA2; PTPN1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1419