Learn More
Biomatik Corporation Phospho-Caspase 3 (Ser150) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51266
Encompass_Preferred
Phospho-Caspase 3 (Ser150) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Caspase 3 (Ser150) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P42574. Gene ID: 836. Target Synonyms: Apopain precursor; CASP-3; CPP-32; CPP32; Caspase-3; Cysteine protease CPP32; ICE3; SCA-1; SREBP cleavage activity 1; Yama protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1438