Learn More
Biomatik Corporation Phospho-CD50/ICAM-3 (Ser518) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51270
Encompass_Preferred
Phospho-CD50/ICAM-3 (Ser518) Colorimetric Cell-Based ELISA Kit for detecting Phospho-CD50/ICAM-3 (Ser518) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P32942. Gene ID: 3385. Target Synonyms: CD50 antigen; CDw50; ICA3; ICAM-R; ICAM3; Intercellular adhesion molecule-3 precursor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1441