Learn More
Biomatik Corporation Phospho-Dematin (Ser403) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51290
Encompass_Preferred
Phospho-Dematin (Ser403) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Dematin (Ser403) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q08495. Gene ID: 2039. Target Synonyms: DEMA; DMT; EPB49; Erythrocyte membrane protein band 4.9. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1458