Learn More
Biomatik Corporation Phospho-EGFR (Tyr1069) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51292
Encompass_Preferred
Phospho-EGFR (Tyr1069) Colorimetric Cell-Based ELISA Kit for detecting Phospho-EGFR (Tyr1069) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P00533. Gene ID: 1956. Target Synonyms: Epidermal growth factor receptor; Proto-oncogene c-ErbB-1; Receptor tyrosine-protein kinase erbB-1; EGFR; ERBB1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1460