Learn More
Biomatik Corporation Phospho-EPHB1/2 (Tyr594/604) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51294
Encompass_Preferred
Phospho-EPHB1/2 (Tyr594/604) Colorimetric Cell-Based ELISA Kit for detecting Phospho-EPHB1/2 (Tyr594/604) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P54762; P29317. Gene ID: 2047; 1969. Target Synonyms: ELK; EPB1; EPHT2; EPTH2; Eph receptor B1; Ephrin type-B receptor 1 precursor; HEK6; NET; Tyrosine-protein kinase receptor EPH-2; kinase EphB1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1462