Learn More
Biomatik Corporation Phospho-HDAC6 (Ser22) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51304
Encompass_Preferred
Phospho-HDAC6 (Ser22) Colorimetric Cell-Based ELISA Kit for detecting Phospho-HDAC6 (Ser22) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q9UBN7. Gene ID: 10013. Target Synonyms: HD6; HDA6; Histone deacetylase 6; JM21. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1471