Learn More
Biomatik Corporation Phospho-AurB (Thr232) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51324
Encompass_Preferred
Phospho-AurB (Thr232) Colorimetric Cell-Based ELISA Kit for detecting Phospho-AurB (Thr232) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q96GD4. Gene ID: 9212. Target Synonyms: AIK2; AIM-1; AIM1; ARK2; AURKB; Aurora- and Ipl1-like midbody-associated protein 1; Aurora-B; Aurora-related kinase 2; Aurora/IPL1-related kinase 2; Serine/threonine protein kinase 12; STK-1; STK12. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1488