Learn More
Biomatik Corporation Phospho-AMPKα (Thr172) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50967
Encompass_Preferred
Phospho-AMPKα (Thr172) Colorimetric Cell-Based ELISA Kit for detecting Phospho-AMPKα (Thr172) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q13131; P54646. Gene ID: 5562; 5563. Target Synonyms: 5'-AMP-activated protein kinase; catalytic alpha-1 chain; AAPK1; AMPK alpha-1 chain; AMPK-alpha1; HMG-CoA redustase kinase; PRKAA1; acetyl-CoA carboxylase kinase; hormone-sensitive lipase kinase; kinase AMPK-alpha1; 5'-AMP-activated protein kinase catalytic subunit alpha-2; AMPK subunit alpha-2; PRKAA2; AMPK; AMPK2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8994