Learn More
Biomatik Corporation Phospho-hnRNP K (Ser284) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51333
Encompass_Preferred
Phospho-hnRNP K (Ser284) Colorimetric Cell-Based ELISA Kit for detecting Phospho-hnRNP K (Ser284) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P61978. Gene ID: 3190. Target Synonyms: 65 kDa phosphoprotein; CSBP; DC-stretch binding protein; Heterogeneous nuclear ribonucleoprotein K; hnRNP-K; HNRNPK; HNRPK; Transformation upregulated nuclear protein; TUNP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1497