Learn More
Biomatik Corporation Phospho-CDC37 (Ser13) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51336
Encompass_Preferred
Phospho-CDC37 (Ser13) Colorimetric Cell-Based ELISA Kit for detecting Phospho-CDC37 (Ser13) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q16543. Gene ID: 11140. Target Synonyms: CC37; Hsp90 chaperone protein kinase-targeting subunit; Hsp90 co-chaperone Cdc37; p50Cdc37. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1500