Learn More
Biomatik Corporation Phospho-Integrin β4 (Tyr1510) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51345
Encompass_Preferred
Phospho-Integrin β4 (Tyr1510) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Integrin β4 (Tyr1510) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P16144. Gene ID: 3691. Target Synonyms: CD104 antigen; GP150; ITB4; integrin beta-4; integrin beta-4C. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1508