Learn More
Biomatik Corporation Phospho-Kir6.2 (Thr224) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51348
Encompass_Preferred
Phospho-Kir6.2 (Thr224) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Kir6.2 (Thr224) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q14654. Gene ID: 3767. Target Synonyms: ATP-sensitive inward rectifier potassium channel 11; IKATP; IRK11; Inward rectifier K channel Kir6.2; KCNJ11; Potassium channel inwardly rectifying subfamily J member 11. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1511